Order Entry
Denmark
ContactUsLinkComponent
Lecithin from egg yolk
Lecithin from egg yolk
Supplier:  TCI
Certificates
Lecithin from egg yolk
Supplier:  TCI
Specification Test Results
  • Appearance:
    Orange to Brown clear liquid to cloudy liquid
  • Saponification Value:
    180.0 to 200.0
  • Acid Value:
    max. 10.0
  • Water:
    max. 2.0 %