
Tetrapentylammonium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4965-17-7
:
Formula:C₂₀H₄₄ClN
MDL Number:MFCD00011857
:334.03 g/mol
Melting Pt:139.85 °C
EINECS:225-606-4
UN:2811
ADR:6.1,I
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together