2-Ethoxyethyl acetate ≥98.0% (by GC)
: TCI
: Ethylglycol acetate
:111-15-9
:
:C₆H₁₂O₃
:MFCD00009251
:156°C
:132.16 g/mol
:–62°C
:55°C
:3 III
:14,03751
:203-839-2
:1172
:GHS02
:GHS07
:GHS08
- Environmentally Preferable:
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together