Order Entry
Denmark
ContactUsLinkComponent
Rifampicin ≥98.0% (by titrimetric analysis)
Irritant
Rifampicin ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Rifampicin ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (HPLC):
    min. 94.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together