Order Entry
Denmark
ContactUsLinkComponent
Ethyl-4-chlorobenzoate ≥96.0% (by GC)
Ethyl-4-chlorobenzoate ≥96.0% (by GC)
Catalog # TCIAC1624-25G
Supplier:  TCI
CAS Number:  
undefined
Ethyl-4-chlorobenzoate ≥96.0% (by GC)
Catalog # TCIAC1624-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :184.62 g/mol
Boiling Pt:238°C
EINECS:230-844-7
CAS Number:7335-27-5
Formula:C₉H₉ClO₂
MDL Number:MFCD00013645