1,10-Phenanthroline monohydrate ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 1,10-Phenanthroline hydrate
CAS Number:5144-89-8
:
Formula:C₁₂H₈N₂·1H₂O
MDL Number:MFCD00149973
:198.22 g/mol
Merck Index:14,07214
Melting Pt:102°C
Density:1,259 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-629-2
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in EtOH (95vol%):almost transparency
- Water:8.6 to 9.6 %
Specification Test Results
Frequently Bought Together