
(R)-(-)-1,3-Butanediol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6290-03-5
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00064277
:90.12 g/mol
Boiling Pt:117°C
Density:1 g/cm³ (20 °C)
Flash Pt:122°C
EINECS:228-532-0
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:-29.0 to -32.0 deg (C=1, EtOH)
Specification Test Results
Frequently Bought Together