Order Entry
Denmark
ContactUsLinkComponent
 
 
3,5-Dihydroxytoluene monohydrate ≥98.0% (by GC)
 :  TCI
 
undefined
 
3,5-Dihydroxytoluene monohydrate ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Solubility in Methanol:
    very faint turbidity
  • Water:
    11.0 to 14.0 %

 

Frequently Bought Together