Boron trifluoride diethyl ether complex ≥98.0% (W)
Supplier: TCI
Certificates
About this item
CAS Number:109-63-7
:
Formula:C₄H₁₀BF₃O
MDL Number:MFCD00013194
:141.93 g/mol
Merck Index:14,01350
Boiling Pt:124°C
Melting Pt:–60°C
Density:1,125 g/cm³ (25 °C)
Flash Pt:59 °C
EINECS:203-689-8
UN:2604
ADR:8,I
:GHS02
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (Gravimetric Method):98.0 to 102.0 %
Specification Test Results
Frequently Bought Together