Order Entry
Denmark
ContactUsLinkComponent
Propionic anhydride ≥98.0% (by GC, titration analysis)
Propionic anhydride ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Propionic anhydride ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Morpholine Method):
    min. 98.0 %

Specification Test Results

Frequently Bought Together