Order Entry
Denmark
ContactUsLinkComponent
 
 
 
Sodium stearate ≥97.0% (by titrimetric analysis)
  TCIAS0081-25G
 :  TCI
 :  
undefined
 
 
Sodium stearate ≥97.0% (by titrimetric analysis)
  TCIAS0081-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 97.0 %

 

 

 :
 :306.46 g/mol
 :14,08678
 :288 °C
 :1,103 g/cm³ (20 °C)
 :Ambient
 :212-490-5
 :3077
 :9,III
 :GHS07
 :GHS09
 :822-16-2
 :C₁₈H₃₅NaO₂
 :MFCD00036404