Order Entry
Denmark
ContactUsLinkComponent
D-(+)-Glucose ≥98.0% (by GC)
D-(+)-Glucose ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
D-(+)-Glucose ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (GC):
    min. 98.0 % (alpha+beta)
  • Specific rotation [a]20/D:
    +50.0 to +54.0 deg (C=10, H2O:NH3 (2+3) = 99.2:0.8)
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together