Propyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
CAS Number:94-13-3
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00002354
Boiling Pt:292
Molecular Weight:180.2 g/mol
Melting Pt:95
Merck Index:14,07866
EINECS:202-307-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:96.0 to 99.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together