4-Bromobenzoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: p-Bromobenzoyl chloride
CAS Number:586-75-4
:
Formula:C₇H₄BrClO
MDL Number:MFCD00000683
:219.46 g/mol
Boiling Pt:247°C
Melting Pt:36 °C
Flash Pt:≥ 110 °C
Storage Temperature:Refrigerator
EINECS:209-580-1
UN:3261
ADR:8,III
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:38.0 to 41.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together