2-Methyl-2-hexanol ≥96.0% (by GC)
: TCI
: 1,1-Dimethyl-1-pentanol, 2-Methylhexan-2-ol
:625-23-0
:
:C₇H₁₆O
:MFCD00004486
:116.2 g/mol
:143°C
:0,8155 g/cm³ (20 °C)
:40°C
:Ambient
:210-881-5
:1987
:3,III
:GHS02
- Environmentally Preferable:
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 96.0 %
Frequently Bought Together