

4-(Trifluoromethyl)phenyl isocyanate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1548-13-6
:
Formula:C₈H₄F₃NO
MDL Number:MFCD00002028
:187.12 g/mol
Boiling Pt:59°C
Melting Pt:58…59 °C
Density:1,31 g/cm³ (20 °C)
Flash Pt:69°C
Storage Temperature:Refrigerator
EINECS:216-284-6
UN:2285
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together