
α,α,α-Trifluoro-p-cresol ≥98.0% (by GC, titration analysis)
: TCI
:402-45-9
:
:C₇H₅F₃O
:MFCD00002363
:162.11 g/mol
:78°C
:45…47 °C
:1,3 g/cm³ (25 °C)
:Refrigerator
:206-945-7
:GHS05
- Appearance:White to Yellow to Orange powder to lump
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:45.0 to 49.0 deg-C
Frequently Bought Together