
4-Ethylresorcinol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2896-60-8
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00002283
:138.17 g/mol
Boiling Pt:155°C
Melting Pt:100 °C
EINECS:220-777-1
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:95.0 to 99.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together