
L(+)-Arginine ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:74-79-3
:
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00002635
:174.2 g/mol
Merck Index:14,00780
Boiling Pt:368 °C (1013 hPa)
Melting Pt:238 °C
Density:1,3 g/cm³ (20 °C)
EINECS:200-811-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Specific rotation [a]20/D:+24.0 to +28.0 deg (C=8, 6mol/L HCl)
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together