Order Entry
Denmark
ContactUsLinkComponent
Benzyl alcohol ≥99.0% (by GC)
Irriterende
Benzyl alcohol ≥99.0% (by GC)
Catalog # TCIAB2378-500G
Leverandør:  TCI
CAS nummer:  
undefined
Irriterende
Benzyl alcohol ≥99.0% (by GC)
Catalog # TCIAB2378-500G
Leverandør:  TCI
CAS nummer:  

Specifikationer

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specifikationstestresultater

Om denne vare

 :
 :108.14 g/mol
Merck Index:14,01124
Kogende Pt:200°C
Smeltende Pt:-15°C
Smeltende Pt:–15°C
Tæthed:1,05 g/cm³ (20 °C)
Flash Pt:100°C
Opbevaringstemperatur:Ambient
EINECS:202-859-9
 :GHS07
CAS nummer:100-51-6
Formel:C₆H₅CH₂OH
MDL nummer:MFCD00004599