1,3-Dihydroxynaphthalene ≥98.0% (by GC, titration analysis)
: TCI
:132-86-5
:
:C₁₀H₈O₂
:MFCD00003965
:160.17 g/mol
:14,06396
:386 °C (1013 hPa)
:124 °C
:1,12 g/cm³ (20 °C)
:205-079-7
:GHS07
:Ambient
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:123.0 to 126.0 deg-C
Frequently Bought Together