
2',6'-Dihydroxyacetophenone ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:699-83-2
:
Formula:C₈H₈O₃
MDL Number:MFCD00002270
:152.15 g/mol
Boiling Pt:296 °C (1013 hPa)
Melting Pt:155…158 °C
Storage Temperature:Ambient
EINECS:211-833-6
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
- Melting point:155.0 to 159.0 deg-C
Specification Test Results
Frequently Bought Together