Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bromophenol blue

Bromophenol blue

Supplier:   TCI
Bromophenol blue
Supplier:  TCI
Certificates
Bromophenol blue

2 Options Available Below

Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:14,01444
  • Melting Pt:273 °C
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Specifications for Product Family

Specification Test Results

  • Appearance:
    Light red to Yellow to Red powder to crystal
  • Absorbance:
    min. 0.480 (5 mg/L, pH 5.0, 590.0 to 594.0 nm)
  • Solubility in Sodium hydroxide solution:
    transparency
  • Solubility in EtOH (50vol%):
    transparency
  • Drying loss:
    max. 1.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 156,00 kr
    156,00 kr
    1 * 1 g
       156,00 kr

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 583,00 kr
    583,00 kr
    1 * 25 g
       583,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.