


Paraformaldehyde ≥90.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:30525-89-4
:
Formula:(CH₂O)n
MDL Number:MFCD00133991
Merck Index:14,07025
Boiling Pt:135 - 136 °C (13 -rr)
Melting Pt:123 °C
Density:1,46 g/cm³ (20 °C)
Flash Pt:70 °C
UN:2213
ADR:4.1,III
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White powder to crystal
- Purity (Sodium Sulfite Method):min. 90.0 %
Specification Test Results
Related Information
Frequently Bought Together