



Hydroxylammonium sulphate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10039-54-0
:
Formula:H₈N₂O₆S
MDL Number:MFCD00044869
:164.14 g/mol
Merck Index:14,04828
Melting Pt:170 °C
Density:1,1 g/cm³ (20 °C)
EINECS:233-118-8
UN:2865
ADR:8,III
:GHS05
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:White powder to crystal
- Purity (Redox method):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together