
2-Nitroresorcinol ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:601-89-8
:
Formula:C₆H₅NO₄
MDL Number:MFCD00007124
:155.11 g/mol
Boiling Pt:234°C
Melting Pt:80…84 °C
Density:1,3526 g/cm³ (96.7 °C)
EINECS:210-010-9
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Brown powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Melting point:83.0 to 86.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together