Order Entry
Denmark
ContactUsLinkComponent
 
Diethanolamine ≥99.0% (by GC) matrix for FABMS and liquid SIMS
 :  TCI
 
undefined
Diethanolamine ≥99.0% (by GC) matrix for FABMS and liquid SIMS
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 99.0 %

 

Frequently Bought Together