


Phenylhydrazinium chloride ≥98.5% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:59-88-1
:
Formula:C₆H₉ClN₂
MDL Number:MFCD00012924
:144.6 g/mol
Merck Index:14,07293
Melting Pt:243…246 °C
EINECS:200-444-7
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Potassium iodate Method):min. 98.5 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together