Order Entry
Denmark
ContactUsLinkComponent
 
D-(-)-Aspartic acid ≥98.0% (by HPLC, titration analysis)
  TCIAA0545-25G
 :  TCI
 :  
undefined
D-(-)-Aspartic acid ≥98.0% (by HPLC, titration analysis)
  TCIAA0545-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White powder to crystal
  • Purity (HPLC) (CAD):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -24.0 to -26.0 deg (C=8, HCl (1+1))
  • Solubility in dilute HCl:
    almost transparency

 

 

 :
 :133.1 g/mol
 :14,00840
 :251 °C
 :217-234-6
 :1783-96-6
 :C₄H₇NO₄
 :MFCD00063081