Order Entry
Denmark
ContactUsLinkComponent
N-phenylanthranilic acid ≥98.0% (by HPLC, titration analysis)
Irritant
N-phenylanthranilic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0873-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
N-phenylanthranilic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0873-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    184.0 to 189.0 deg-C

Specification Test Results

About this item

 :
 :213.24 g/mol
Merck Index:14,07273
Boiling Pt:375 °C (1013 hPa)
Melting Pt:182…185 °C
EINECS:202-066-8
 :GHS07
CAS Number:91-40-7
Formula:C₁₃H₁₁NO₂
MDL Number:MFCD00002421