
Copper(I) acetate ≥93.0% (by titrimetric analysis)
: TCI
:598-54-9
:
:C₂H₃CuO₂
:MFCD00058908
:122.59 g/mol
:14,02658
:250 °C
:209-938-7
:GHS07
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (Potassium permanganate method):min. 93.0 %
Frequently Bought Together