Spironolactone ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:52-01-7
:
Formula:C₂₄H₃₂O₄S
MDL Number:MFCD00082250
Merck Index:14,08760
Boiling Pt:597 °C (1013 hPa)
Melting Pt:196 °C
:416.58 g/mol
EINECS:200-133-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (UV-vis method):min. 95.0 %
- Specific rotation [a]20/D:-34.0 to -38.0 deg (C=1, CHCl3)
- Solubility in EtOH:very faint turbidity
Specification Test Results
Frequently Bought Together