Order Entry
Denmark
ContactUsLinkComponent
2-Methyl-1,3-cyclohexanedione ≥98.0% (by GC, titration analysis)
2-Methyl-1,3-cyclohexanedione ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
2-Methyl-1,3-cyclohexanedione ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together