Order Entry
Denmark
ContactUsLinkComponent
 
2-Indolinone ≥98.0% (by GC)
 :  TCI
 
undefined
2-Indolinone ≥98.0% (by GC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    125.0 to 130.0 deg-C

 

Frequently Bought Together