
1-Bromo-4-methylpentane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:626-88-0
:
Formula:C₆H₁₃Br
MDL Number:MFCD00013544
Boiling Pt:148°C
Melting Pt:–71 °C
Density:1,134 g/cm³ (25 °C)
Flash Pt:57°C
EINECS:210-968-8
ADR:3,III
:165.07 g/mol
UN:1993
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together