PIG® sump skimmers
: NEW PIG
Keep sumps, tanks and bilges oil-free with easy-to-retrieve PIG Sump Skimmers.
- Simply lower this floating skimmer into any water-based liquid to absorb oils without taking in water
- Ideal for soaking up unwanted oil-based fluids that collect in sumps, tanks and bilges
- Polypropylene loop can be attached to rope (not included) for easy deployment and retrieval
- Absorbs and retains oils and oil-based liquids - including lubricants and fuels - without taking in a drop of water
- Bright white colour makes absorbed oil easier to see; clearly shows saturation level
Can be incinerated after use to reduce waste or for fuels blending. Polypropylene skin resists chemicals and tearing; meets NFPA 99 standards for static decay.
Frequently Bought Together