Skimmer pillows, PIG® Oil-Only
Supplier: NEW PIG
Certificates
About this item
The polypropylene filler is chemical resistant and hydrophobic for containing and absorbing oil-based spills in a variety of environments.
- High capacity absorbent to quickly soak up liquids
- Pillows float to clean up spills on water confined by booms or socks
- Pillows repel rainwater and are UV resistant, making them ideal for outdoor applications
Frequently Bought Together