Order Entry
Denmark
ContactUsLinkComponent
Test tube racks, wire
Test tube racks, wire
Catalog # HECH42815248
undefined
Test tube racks, wire
Catalog # HECH42815248
Specifications
  • For tube Ø:
    12 mm
  • No. of Holes:
    48 (4×12)
  • Colour:
    White
  • Height:
    40 mm
  • Pk:
    1
Specifications
About this item
Racks, made of plastic coated wire, ideally used for test tubes or centrifuge tubes.
  • Good chemical resistance
  • Good temperature resistance