Order Entry
Denmark
ContactUsLinkComponent
 
4-(4-Nitrophenylazo)resorcinol for synthesis, Sigma-Aldrich®
  8.41713.0025
 :  Merck
 :  
4-(4-Nitrophenylazo)resorcinol for synthesis, Sigma-Aldrich®
  8.41713.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    183 °C (decomposition)
  • Molar Mass:
    259.22 g/mol

 

 

 :
 :259.22 g/mol
 :M-Clarity™ quality level = MQ200
 :200-808-5
 :74-39-5
 :C₁₂H₉N₃O₄
 :MFCD00007310