Glycerol monostearate, purified
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31566-31-1
:
Formula:C₂₁H₄₄O₅
MDL Number:MFCD00036186
:376.57 g/mol
Merck Index:14,4489
Storage Temperature:Ambient
EINECS:250-705-4
Related Information
Frequently Bought Together