Organic reference standard, Tetrabromobisphenol A 50 µg/ml in methanol
Catalog # CAMBULM-4694-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:79-94-7
- Description:Tetrabromobisphenol A 50 µg/ml in methanol
- Formula:C₁₅H₁₂Br₄O₂
- Description:Tetrabromobisphenol A