Order Entry
Denmark
ContactUsLinkComponent
Methyl D-glucopyranoside (mixture of anomers) for synthesis, Sigma-Aldrich®
Methyl D-glucopyranoside (mixture of anomers) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Methyl D-glucopyranoside (mixture of anomers) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    600 g/l (25 °C)
  • Melting Point:
    155 - 165 °C
  • Molar Mass:
    194.18 g/mol
  • Bulk Density:
    400 kg/m3

Specification Test Results

Related Information

Frequently Bought Together