Order Entry
Denmark
ContactUsLinkComponent
Desiccators, NOVUS
Desiccators, NOVUS
Catalog # WITG5.660.200
Supplier:  witeg Labortechnik
undefined
Desiccators, NOVUS
Catalog # WITG5.660.200
Supplier:  witeg Labortechnik
Specifications
  • Ø int.:
    200 mm
  • Type:
    NOVUS, with needle valve stopcock, cone ST 24/29
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This desiccator is made from DURAN® glass, without porcelain disc, with tubulated lid.