Integrated vertical electrophoresis unit, SE300 miniVE™
SE300 miniVE™ integrated vertical electrophoresis and blotting unit allows performance of both electrophoresis and electro transfer in one sturdy, compact unit.
- Quick, excellent results using minimal buffer
- Passive cooling produces gels with straight lanes and sharp bands
- Transfer up to 4 gels in 45 minutes with only 300 ml buffer using optional SE302 blot modules
- Designed for use with a wide variety of precast gels
: Complete system includes lower buffer chamber (tank), gel modules with gaskets (2 assemblies), safety lid with high voltage leads, 3× rectangular glass plates (10×10,5 cm), 3× notched glass plates (10×10,5 cm), combs (2× 10-well, 1,0 mm thick), 4× T-Spacers, (1,0 mm thick), 2× well locating decals, plate separation tool, and gel seal. Basic unit includes same as above without the combs, spacers and glass plates, which can be ordered separately.
- Catalog No:
- 732-1772
- 732-1773
- Model:
- SE300
- SE300-10A-1.0
- Glass plate size (W×H):
- 100×105 or 100×80 mm
- 100×105 or 100×80 mm
- Humidity:
- < 80% RH
- < 80% RH
- Max. temperature:
- 45 °C
- 45 °C
- Unit W×H×D:
- 192×172×188 mm
- 192×172×188 mm
Blot Module
Frequently Bought Together