Order Entry
Denmark
ContactUsLinkComponent
Sleeve protectors, Aracut® Sleeves II
Sleeve protectors, Aracut® Sleeves II
  112-2918
undefined
Sleeve protectors, Aracut® Sleeves II
  112-2918
  • Garment application:
    Industrial,
    Mechanical protection
  • Gender:
    Unisex
  • Risk area:
    Heat,
    Cuts
  • Colour:
    Yellow
  • Length:
    360 mm
  • Fabric:
    Kevlar® fiber with leather reinforcement
  • Pk:
    1
This Kevlar® knitted sleeve with thumbhole and leather reinforcement is ideal for handling sharp and hot objects in a dry atmosphere.
  • Kevlar® fibre offers good resistance to cuts and protection against thermal hazards (contact heat)
  • Reinforced thumb and index finger offer greater cuff support
  • Split cowhide leather reinforcement helps to improve protection of the forearm
  • Dual thickness (2 ply Kevlar®) improves the cuff's mechanical and thermal performance
Suitable industries are the glass and paper industry, as well as automotive and car equipment manufacturers.
Gauge: 10
 : EN 388 (4443), EN 407 (X2XXXX); Cat. II