L-Methionine-[methyl-¹³C] (99% ¹³C )
Catalog # CAMBCLM-206-PK
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1
- Seq:0001
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:49705-26-2
Formula:C₅H₁₁NO₂S