
7-Amino-4-(trifluoromethyl)coumarin ≥99%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:53518-15-3
:
Formula:C₁₀H₆F₃NO₂
MDL Number:MFCD00006858
:229.16 g/mol
Melting Pt:221...222 °C (lit.)
:M-Clarity™ quality level = MQ100
EINECS:258-599-1
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Color):Yellow
- Appearance (Form):Powder or Crystals or Chunk(s) or Powder with Chunk(s) or Crystals with Chunk(s)
- Infrared spectrum:Conforms to Structure
- Carbon:51.9 - 52.9 %
- Nitrogen:5.9 - 6.3 %
- Purity (GC):≥99.0 %
Specification Test Results
Frequently Bought Together