3-Methyl-1-phenyl-2-pyrazolin-5-one
Catalog # CAYM13320-500
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:174.2 g/mol
EINECS:201-891-0
CAS Number:89-25-8
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD00003138