D-(-)-N-Methylglucamine
Glass bottle for solids, 10 mg
Catalog # EDQMY0001209
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
- CAS Number:6284-40-8
- Formula:H₃CNHCH₂(CH(OH))₄CH₂OH
- MDL Number:MFCD00004707
- Molecular Weight:195.22 g/mol
- Merck Index:12,06154
- Melting Pt:128…131 °C
- Storage Temperature:Ambient
- EINECS:228-506-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



