Glycine
12 kg
Supplier: CHATTEM CHEMICALS
Jump to:
Product Details & Documents
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:13,04504
- Boiling Pt:181 °C (1013 hPa)
- Melting Pt:83 °C
- Density:1.64 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-272-2
Documents
Specifications
Specifications
- Pk:12 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



